Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.
Thermo Scientific™

MAP3K7/MAP3K7IP1 (TAK1-TAB1) Recombinant Human Protein

Catalog number: PR8004B
Thermo Scientific™

MAP3K7/MAP3K7IP1 (TAK1-TAB1) Recombinant Human Protein

Catalog number: PR8004B
Catalog Number
PR8004B
Unit Size
100 µg
Price (USD)
Price: 1,646.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 6,980.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 486.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PR8004B100 µg
Price: 1,646.00
Your Price:
-
PV44301 mg
Price: 6,980.00
Your Price:
-
PV439410 µg
Price: 486.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
MAP3K7 (TAK1) controls transcription regulation and apoptosis, and is implicated in inflammatory disease and rheumatoid arthritis. Fusion of MAP3K7 to MAP3K7IP1 (TAB1) activates MAP3K7.

Specifications

Accession Number
NP_663306, NP_006107
Molecular Weight
45.8 kDa
Quantity
100 µg
Gene Alias
MAP3K7/MAP3K7IP1 (TAK1-TAB1)
KinaseFamily
MLK Kinase Family
Recombinant
Yes
Protein Tag
His Tag
Kinase Group
Ser⁄Thr Kinases: TKL Group
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Form
Full Length
Form
Frozen
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.