Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.
Thermo Scientific™

MET (cMET) Recombinant Human Protein

Catalog number: PV3588
Thermo Scientific™

MET (cMET) Recombinant Human Protein

Catalog number: PV3588
Catalog Number
PV3588
Unit Size
1 mg
Price (USD)
Price: 7,110.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,674.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 485.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV35881 mg
Price: 7,110.00
Your Price:
-
PR4563B100 µg
Price: 1,674.00
Your Price:
-
PV314310 µg
Price: 485.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
MET (cMET) is a receptor-like tyrosine kinase whose disregulation has been linked to many types of human malignancies.

Specifications

Accession Number
NP_000236.2
Molecular Weight
53.7 kDa
Quantity
1 mg
Gene Alias
MET (cMET)
KinaseFamily
MET (HGFR) Kinase Family
Recombinant
Yes
Protein Tag
His Tag
Kinase Group
Tyrosine Kinases
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Form
Cytoplasmic
Form
Frozen
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 20 mM Tris, pH 7.5, 100 mM NaCl, 0.5 mM EDTA, 0.05% Triton X-100, 2 mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.