Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.
Thermo Scientific™

PRKCQ (PKC theta) Recombinant Human Protein

Catalog number: PV3605
Thermo Scientific™

PRKCQ (PKC theta) Recombinant Human Protein

Catalog number: PV3605
Catalog Number
PV3605
Unit Size
1 mg
Price (USD)
Price: 6,930.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
5 µg
Price (USD)
Price: 392.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,646.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV36051 mg
Price: 6,930.00
Your Price:
-
P29965 µg
Price: 392.00
Your Price:
-
PR4143B100 µg
Price: 1,646.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The PKC family of serine⁄threonine kinases, including PRKCQ (PKC theta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.

Specifications

Accession Number
NP_006248
Molecular Weight
84.4 kDa
Purity
90% by SDS-PAGE
Quantity
1 mg
Gene Alias
PRKCQ (PKC theta)
KinaseFamily
PKC Kinase Family
Recombinant
Yes
Protein Tag
His Tag
Kinase Group
Ser⁄Thr Kinases: AGC Group
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Form
Full Length
Form
Frozen
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 20mM HEPES, pH 7.5, 150mM NaCl, 0.5mM EGTA, 0.5mM EDTA, 0.05% NP-40, 5mM DTT, 20% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.