Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.
Thermo Scientific™

CDK5/p35 Recombinant Human Protein

Catalog number: PV3981
Thermo Scientific™

CDK5/p35 Recombinant Human Protein

Catalog number: PV3981
Catalog Number
PV3981
Unit Size
1 mg
Price (USD)
Price: 7,010.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 477.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,646.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV39811 mg
Price: 7,010.00
Your Price:
-
PV300010 µg
Price: 477.00
Your Price:
-
PR4026C100 µg
Price: 1,646.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
CDK5 is a cyclin-dependent kinase that plays a critical role in neuron development. The kinase is strongly activated when it complexes with neuron-specific activators, especially p35, but less strongly when it complexes with cyclins. This protein is coexpressed and copurified with p35.

Specifications

Accession Number
NP_004926.1 I NP_003876
Molecular Weight
74.5 kDa
Quantity
1 mg
Gene Alias
CDK5/p35
KinaseFamily
CDK and CHK Kinase Family
Recombinant
Yes
Protein Tag
His Tag
Kinase Group
Ser⁄Thr Kinases: CMCG Group
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Form
Full Length
Form
Frozen
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 20% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.