Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.
Thermo Scientific™

CSNK2A2 (CK2 α 2) Recombinant Human Protein

Catalog number: PV3993
Thermo Scientific™

CSNK2A2 (CK2 α 2) Recombinant Human Protein

Catalog number: PV3993
Catalog Number
PV3993
Unit Size
1 mg
Price (USD)
Price: 7,010.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,650.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 479.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV39931 mg
Price: 7,010.00
Your Price:
-
PR6299B100 µg
Price: 1,650.00
Your Price:
-
PV362410 µg
Price: 479.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
Casein kinase II is a protein serine⁄threonine kinase that is thought to be present in all eukarotic cells, implying that it has fundamental cellular functions. The holoenzyme is a tetramer containing 2 alpha or alpha-prime subunits (or one of each) and 2 beta subunits. The function of beta subunits is unknown and the alpha subunit is the catalytic unit.

Specifications

Accession Number
NP_001887
Molecular Weight
69.2 kDa
Quantity
1 mg
Gene Alias
CSNK2A2 (CK2 α 2)
KinaseFamily
CK2 Kinase Family
Recombinant
Yes
Protein Tag
GST Tag
Kinase Group
Ser⁄Thr Kinases: CK1 Group
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Form
Full Length
Form
Frozen
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.