Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.
Thermo Scientific™

RPS6KA1 (RSK1) Recombinant Human Protein

Catalog number: PV4049
Thermo Scientific™

RPS6KA1 (RSK1) Recombinant Human Protein

Catalog number: PV4049
Catalog Number
PV4049
Unit Size
1 mg
Price (USD)
Price: 7,010.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,644.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 480.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV40491 mg
Price: 7,010.00
Your Price:
-
PR6812A100 µg
Price: 1,644.00
Your Price:
-
PV368010 µg
Price: 480.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
RPS6KA1 (RSK1) is a growth-factor regulated serine⁄threonine kinase that is involved in the MAPK cascade. It is thought to control cellular proliferation and differentiation through the phosphorylation of transcription factors.

Specifications

Accession Number
NP_002944
Molecular Weight
88.5 kDa
Quantity
1 mg
Gene Alias
RPS6KA1 (RSK1)
KinaseFamily
RSK (RPS6K) Kinase Family
Recombinant
Yes
Protein Tag
His Tag
Kinase Group
Ser⁄Thr Kinases: AGC Group
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Form
Full Length
Form
Frozen
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.