Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.
Thermo Scientific™

MERTK (cMER) Recombinant Human Protein

Catalog number: PV4112
Thermo Scientific™

MERTK (cMER) Recombinant Human Protein

Catalog number: PV4112
Catalog Number
PV4112
Unit Size
1 mg
Price (USD)
Price: 7,010.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,644.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 480.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV41121 mg
Price: 7,010.00
Your Price:
-
PR6317A100 µg
Price: 1,644.00
Your Price:
-
PV362710 µg
Price: 480.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
MERTK (cMER) is a tyrosine kinase proto-oncogene and is involved in the retinal pigment epithelium (RPE) phagocytosis pathway, which is implicated in human retinal disease.

Specifications

Accession Number
NP_006334
Molecular Weight
63.4 kDa
Quantity
1 mg
Gene Alias
MERTK (cMER), MER
KinaseFamily
AXL Kinase Family
Recombinant
Yes
Protein Tag
GST Tag
Kinase Group
Tyrosine Kinases
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Form
Cytoplasmic
Form
Frozen
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.