Thermo Scientific™

ALK Recombinant Human Protein

Catalog number: PV4185
Thermo Scientific™

ALK Recombinant Human Protein

Catalog number: PV4185
Catalog Number
PV4185
Unit Size
1 mg
Price (USD)
Price: 7,380.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 481.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,650.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV41851 mg
Price: 7,380.00
Your Price:
-
PV386710 µg
Price: 481.00
Your Price:
-
PR7396B100 µg
Price: 1,650.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
Anaplastic Lymphoma Receptor Tyrosine Kinase (ALK) belongs to the insulin receptor superfamily. It is vital for brain development. Mutations, rearrangements, and amplifications in the ALK gene have been found in tumors, including anaplastic large cell lymphoma, neurblastoma, and non-small cell lung cancer. Chromosomal rearrangement is the most common genetic alteration.

Learn more about our kinase proteins >

Related Links:

LanthaScreen® Kinase Activity Assay
LanthaScreen® Eu Kinase Binding Assay

Specifications

Accession Number
NP_004295
Gene ID (Entrez)
238
Molecular Weight
89.9 kDa
Quantity
1 mg
Gene Alias
CD246, NBLST3
KinaseFamily
ALK Kinase Family
Recombinant
Yes
Protein Tag
GST Tag
Kinase Group
Receptor Tyrosine Kinase
Expression System
Baculovirus
Protein Form
Active
Form
Liquid
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol

Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.