Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com to ensure your needs are accommodated

88 more kinases now available to order! See the complete list.
Catalog Number
PV4213
Unit Size
10 µg
Price (USD)
Price: 481.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,646.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 7,010.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV421310 µg
Price: 481.00
Your Price:
-
PR7829A100 µg
Price: 1,646.00
Your Price:
-
PV43251 mg
Price: 7,010.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
RIPK2 is a serine⁄threonine kinase that is involved in cell activation, proliferation, differentiation, and apoptosis.

Specifications

Accession Number
NP_003812
Molecular Weight
38.9 kDa
Quantity
10 µg
Gene Alias
RIPK2, RICK, RIP2
KinaseFamily
RIPK Kinase Family
Protein Length
aa 1-299
Recombinant
Yes
Protein Tag
His Tag
Assay Entry
Biochemical binding/kinetic
Kinase Group
Ser⁄Thr Kinases: TKL Group
Shipping Condition
Dry Ice
System Compatibility
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Target Entry
RIPK2
Target Gene
receptor-interacting serine-threonine kinase 2
Expression System
Baculovirus
Protein Family
Kinases & Inhibitors
Protein Form
Recombinant, Catalytic
Form
Liquid
Protein Subtype
Serine⁄Threonine Kinases
Validated Application
Kinase Assay
Species
Human
Research Category
Signal Transduction, Apoptosis

Contents & Storage

Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.