Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com to ensure your needs are accommodated

88 more kinases now available to order! See the complete list.
Thermo Scientific™

PI4KA Human Protein

Catalog number: PV5689
Thermo Scientific™

PI4KA Human Protein

Catalog number: PV5689
Catalog Number
PV5689
Unit Size
20 µg
Price (USD)
Price: 780.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,646.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 7,010.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV568920 µg
Price: 780.00
Your Price:
-
PR9056A100 µg
Price: 1,646.00
Your Price:
-
PV56901 mg
Price: 7,010.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
PI4KB and PI4KA are the two human type III PI4 kinases, which share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol.

Specifications

Accession Number
NP_002641.1
Gene ID (Entrez)
PI4KA
Molecular Weight
124.0 kDa
Quantity
20 µg
Gene Alias
PI4KA
KinaseFamily
PI3 Kinase Family
Protein Length
Full-length
Protein Tag
GST Tag
Assay Entry
Biochemical ADP accumulation
Kinase Group
Lipid Kinases
Shipping Condition
Dry Ice
System Compatibility
Adapta™
Tag Position
N-Terminal
Target Entry
PI4KA
Target Gene
phosphatidylinositol 4-kinase, catalytic, alpha, PI4KA
Expression System
Baculovirus
Protein Family
Kinases & Inhibitors
Protein Form
Full Length, Full
Form
Frozen
Protein Subtype
Lipid Kinases
Validated Application
Kinase Assay
Species
Human
Research Category
Signal Transduction

Contents & Storage

50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws