Informational:Currently, the 1 mg pack size of DNA PK does not include DNA-PK Activator (PV5884)—to obtain activator, please contact us at discoverysciences@lifetech.com.
——————————————————————————————————————————————————————————————————
Informational:For order quantities > 1, multiple lots may be used to fulfill the order. If you require a single lot, please email discoverysciences@lifetech.com.

During checkout, this product will be reported as "Out of Stock" with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.

88 more kinases now available to order! See the complete list.
Informational:During checkout, this product will be reported as “Out of Stock” with an estimated availability date. Often, the actual availability date will be earlier. Send inquiries regarding actual availability date to discoverysciences@lifetech.com.
——————————————————————————————————————————————————————————————————
Thermo Scientific™

DNA-PK Human Protein

Catalog number: PV5865
Thermo Scientific™

DNA-PK Human Protein

Catalog number: PV5865
Catalog Number
PV5865
Unit Size
1 mg
Price (USD)
Price: 12,000.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,658.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 482.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV58651 mg
Price: 12,000.00
Your Price:
-
PR9107A100 µg
Price: 1,658.00
Your Price:
-
PV586610 µg
Price: 482.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
DNA-PK is a heterotrimeric, DNA-dependent protein kinase that initiates signalling cascades in response to DNA damage. DNA-PK is an atypical kinase, closely related to the PI3 kinases, and is often potently inhibited by small molecules designed to target PI3 kinases.

Specifications

Accession Number
0.0
Molecular Weight
600 kDa
Quantity
1 mg
Gene Alias
DNA-PK
KinaseFamily
PIKK Kinase Family
Protein Tag
Untagged
Kinase Group
Ser⁄Thr Kinases: Atypical
Shipping Condition
Dry Ice
Expression System
Baculovirus
Protein Family
Kinases & Inhibitors
Form
Frozen
Validated Application
Kinase Assay
Species
Human
Research Category
Drug Development

Contents & Storage

Kinase proteins are shipped on dry ice and should be stored at -80°C.