Thermo Scientific™

ALK [R1275Q] Recombinant Human Protein

Catalog number: PV6171
Thermo Scientific™

ALK [R1275Q] Recombinant Human Protein

Catalog number: PV6171
Catalog Number
PV6171
Unit Size
1 mg
Price (USD)
Price: 7,380.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 478.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
100 µg
Price (USD)
Price: 1,650.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV61711 mg
Price: 7,380.00
Your Price:
-
PV616910 µg
Price: 478.00
Your Price:
-
PV6170100 µg
Price: 1,650.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
Human ALK R1275Q, internal fragment, amino acids L1066-S1437 (as in NCBI/Protein entry NP_004295.2), R1275Q mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.

• Validated in LanthaScreen® Kinase Binding Assay
• Protocols available for simple use and optimized performance
• Multiple pack sizes available for convenient use

Related Product:
LanthaScreen® Eu Kinase Binding Assay

Specifications

Molecular Weight
70.4 kDa
Quantity
1 mg
Protein Length
aa 1066-1437
Recombinant
Yes
Protein Tag
GST Tag, His Tag
Kinase Group
Receptor Tyrosine Kinase
Expression System
Insect
Protein Form
Recombinant
Form
Liquid
Validated Application
Kinase Assay
Species
Human

Contents & Storage

Storage Buffer: 50 mM HEPES pH 7.5, 100 mM NaCl, 5 mM DTT, 15 mM reduced glutathione, 20% glycerol

Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.