Thermo Scientific™

CDK11 Recombinant Human Protein, Inactive

Catalog number: PV6284
Thermo Scientific™

CDK11 Recombinant Human Protein, Inactive

Catalog number: PV6284
Catalog Number
PV6284
Unit Size
100 µg
Price (USD)
Price: 1,650.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
10 µg
Price (USD)
Price: 481.00
Your Price:
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Price: 7,370.00
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV6284100 µg
Price: 1,650.00
Your Price:
-
PV628310 µg
Price: 481.00
Your Price:
-
PV62851 mg
Price: 7,370.00
Your Price:
-
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
CDK11 is a member of the cyclin-dependent kinase 11 family which is known to be essential for eukaryotic cell cycle control and it is in close proximity to CDC2L2, a nearly identical gene in the same chromosomal region. CDK11 is a negative regulator of normal cell cycle progression and Its expression is regulated during murine embryogenesis, and its activity is coordinately regulated with that of p34 (cdc2) during the cell cycle. CDK11 gene may harbor 1 or more tumor suppressor genes that affected by chromosome 1p36 modifications in neuroblastoma. CDK11 could be cleaved by caspases and was demonstrated to play roles in cell apoptosis. Recombinant full-length human CDK11 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.

• Validated in LanthaScreen® Kinase Binding Assay
• Protocols available for simple use and optimized performance
• Multiple pack sizes available for convenient use

Related Product:
LanthaScreen® Eu Kinase Binding Assay

Specifications

Molecular Weight
84.2 kDa
Quantity
100 µg
Recombinant
Yes
Protein Tag
GST Tag
Kinase Group
Serine/Threonine Kinase
Expression System
Baculovirus
Protein Family
Kinases & Inhibitors
Protein Form
Recombinant
Form
Liquid
Validated Application
Kinase Assay
Species
Human

Contents & Storage

Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol

Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.