You need to enable JavaScript to run this app.
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
Home
Shop All Products
Diagnostic Tests and Controls
Chemistry Controls, Calibrators and Reagents
General Microbiology Quality Control Products
BactiDisks™ Corynebacterium diphtheriae ATCC™ 13812™
Thermo Scientific™
BactiDisks™ Corynebacterium diphtheriae ATCC™ 13812™
Catalog number:
R19015
Related applications:
Clinical Microbiology
Technical Support
Customer Service
Thermo Scientific™
BactiDisks™ Corynebacterium diphtheriae ATCC™ 13812™
Catalog number:
R19015
Related applications:
Clinical Microbiology
Technical Support
Customer Service
Catalog Number
R19015
Unit Size
-
Price (USD)
Request A Quote
Save to list
Catalog Number
Unit Size
Price (USD)
R19015
-
Request A Quote
Save to list
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
Specifications
Description
Corynebacterium diphtheriae
Product Type
Organism
Target Organism Class
Gram-Positive Rods
Quantity
10 Disk/Vial
Certifications/Compliance
Culti-Loops are CE Marked.
ATCC®
13812™*
Test Type
Qualitative QC
Type
Single Organism
Format
BactiDisks