UriSponge™ Urine Specimen Collection, Preservation and Transport System
Catalog number: R723401
UriSponge™ Urine Specimen Collection, Preservation and Transport System
Catalog number: R723401
Simplify the collection and preservation of urine specimens without added expense and hazard of hypodermic urine transfer straws using COPAN™ UriSponge™ Urine Specimen Collection, Preservation and Transport System.
As a part of the Liquid Based Microbiology product line, the UriSponge™ Urine Specimen Collection, Preservation, and Transport System combines a plastic tube and a screw cap with attached sponges impregnated with boric acid and sodium formate urine preservatives. Simply collect a sample by dipping the sponge momentarily into the urine specimen.
Catalog Number
R723401
Unit Size
Each
Item Description
Bulk (12x80) 500/case
Price (USD)
Full specifications
Product Type SpecsUrine Specimen Collection, Preservation and Transport System
DescriptionUriSponge™ Urine Specimen Collection, Preservation and Transport System
Item DescriptionBulk (12x80) 500/case
Unit SizeEach
Catalog Number
Unit Size
Each
Item Description
Pouch (16x100) 300/case
Price (USD)
Full specifications
Product Type SpecsUrine Specimen Collection, Preservation and Transport System
DescriptionUriSponge™ Urine Specimen Collection, Preservation and Transport System
Item DescriptionPouch (16x100) 300/case
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeItem DescriptionPrice (USD)
R723401Full specifications
EachBulk (12x80) 500/caseRequest A Quote
Product Type SpecsUrine Specimen Collection, Preservation and Transport System
DescriptionUriSponge™ Urine Specimen Collection, Preservation and Transport System
Item DescriptionBulk (12x80) 500/case
Unit SizeEach
R723400Full specifications
EachPouch (16x100) 300/caseRequest A Quote
Product Type SpecsUrine Specimen Collection, Preservation and Transport System
DescriptionUriSponge™ Urine Specimen Collection, Preservation and Transport System
Item DescriptionPouch (16x100) 300/case
Unit SizeEach
Showing 2 of 2
Eliminate the need to adhere to precise fill lines and reduce the bacterial inhibition from overexposure to preservatives or overgrowth associated with traditional vacuum boric acid systems with UriSponge Urine Specimen Collection, Preservation and Transport System by maintaining the urine-to-preservative ratio. Preservatives are evenly dispersed within the sponge applicator of the UriSponge and are only activated upon contact with urine.
 UriSponge™ offers a pleasant clinical experience and exceptional laboratory value:
  • Automatically processed on WASP® using its built-in spinner, which easily elutes urine from the sponges prior to planting and streaking. If processed manually, simply centrifuge the tube to release urine from the sponge.
  • No hypodermic needle urine transfer straw needed, eliminating potential injury and sharps disposal costs 
  • Reduce repeat collections from patient due to inaccurate filling of boric acid tubes
  • Maintains viability of bacteria at refrigerated and room temperature for up to 48 hours
For In Vitro Diagnostic Use.