Thermo Scientific™

Screw Cap Septum Vials

Catalog number: TS-13080-CMD
Thermo Scientific™

Screw Cap Septum Vials

Catalog number: TS-13080-CMD
For a convenient, versatile, and cost-effective vial and closure system, choose Thermo Scientific™ Screw Cap Septum Vials. Made of autoclavable borosilicate glass and available with a wide assortment of special closures and accessories, these vials are ideal for sample collection and small derivatization reactions for use with automated gas (GC) and liquid chromatography (LC), and for storage of reagents and standards under complete seal with instant syringe access. The heavy-duty, flip-top divider box provides easy access to vials, caps, and septa and offers a convenient sample storage center. Clear and amber vials are available.
 
Catalog Number
TS-13080-CMD
Unit Size
Each
Volume (Metric)
1.5 mL
Price (USD)
Full specifications
Description1.5mL; Amber
ColorAmber
Diameter (Metric) Outer12 mm
MaterialBorosilicate Glass
Volume (Metric)1.5 mL
Volume (Metric) Total1.5 mL
Vial TypeSeptum
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)32 mm
Certifications/ComplianceU.S. EPA 40 CFR
TypeVial
Closure Size8-425
Unit SizeEach
Catalog Number
Unit Size
Pack of 72
Volume (Metric)
3.5 mL
Price (USD)
Full specifications
Description3.5mL; Clear
ColorClear
Diameter (Metric) Outer15 mm
MaterialBorosilicate Glass
Volume (Metric)3.5 mL
Volume (Metric) Total3.5 mL
Vial TypeSeptum
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)45 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeVial
Closure Size13-425
Unit SizePack of 72
Catalog Number
Unit Size
Pack of 72
Volume (Metric)
7 mL
Price (USD)
Full specifications
Description7mL; Clear
ColorClear
Diameter (Metric) Outer17 mm
MaterialBorosilicate Glass
Volume (Metric)7 mL
Volume (Metric) Total7 mL
Vial TypeSeptum
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)60 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeVial
Closure Size15-425
Unit SizePack of 72
Catalog Number
Unit Size
-
Volume (Metric)
14 mL
Price (USD)
Full specifications
Description14mL; Clear
ColorClear
Diameter (Metric) Outer21 mm
MaterialBorosilicate Glass
Volume (Metric)14 mL
Volume (Metric) Total14 mL
Vial TypeSeptum
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)70 mm
Certifications/ComplianceU.S. EPA 40 CFR Part 136, 141
TypeVial
Closure Size18-400
Unit Size-
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeVolume (Metric)Price (USD)
TS-13080-CMDFull specifications
Each1.5 mLRequest A Quote
Description1.5mL; Amber
ColorAmber
Diameter (Metric) Outer12 mm
MaterialBorosilicate Glass
Volume (Metric)1.5 mL
Volume (Metric) Total1.5 mL
Vial TypeSeptum
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)32 mm
Certifications/ComplianceU.S. EPA 40 CFR
TypeVial
Closure Size8-425
Unit SizeEach
TS-13019Full specifications
Pack of 723.5 mLContact Us ›
Description3.5mL; Clear
ColorClear
Diameter (Metric) Outer15 mm
MaterialBorosilicate Glass
Volume (Metric)3.5 mL
Volume (Metric) Total3.5 mL
Vial TypeSeptum
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)45 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeVial
Closure Size13-425
Unit SizePack of 72
TS-13028Full specifications
Pack of 727 mLContact Us ›
Description7mL; Clear
ColorClear
Diameter (Metric) Outer17 mm
MaterialBorosilicate Glass
Volume (Metric)7 mL
Volume (Metric) Total7 mL
Vial TypeSeptum
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)60 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeVial
Closure Size15-425
Unit SizePack of 72
TS-13043Full specifications
-14 mLRequest A Quote
Description14mL; Clear
ColorClear
Diameter (Metric) Outer21 mm
MaterialBorosilicate Glass
Volume (Metric)14 mL
Volume (Metric) Total14 mL
Vial TypeSeptum
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)70 mm
Certifications/ComplianceU.S. EPA 40 CFR Part 136, 141
TypeVial
Closure Size18-400
Unit Size-
Showing 4 of 4
Applications

The 40mL clear or amber vial with a Thermo Scientific Disc is suitable for discrete water sampling under U.S. EPA 40 CFR Parts 136 and 141.

The 1.5mL clear and amber vials fit GC and LC autosamplers that use standard 12 × 32 mm vials.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.