Extend the life of the quarry
The first step in the cement process is the recovery of the limestone and sometimes the extraction of clay. When unwanted elements reduce the mineable areas of the quarry, the CB Omni Fusion Online Elemental Analyzer, using PGNAA technology, can monitor material chemistry in real time and allow for immediate adjustments. Other Thermo Scientific solutions include Ramsey Belt Scale Systems to measure the amount of materials that is being mined, and the Oretronic Tramp Metal Detectors that monitor for unwanted metals in the materials. Thermo Scientific handheld XRF analyzers can quickly analyze samples from the quarry.
Learn more about the Cement Production Process
Achieve consistent stockpile control and material sorting and blending. The Thermo Scientific CB Omni Agile provides real-time online elemental analysis of an entire raw material process stream using a choice of Prompt Gamma Neutron Activation Analysis (PGNAA) or Pulsed Fast Thermal Neutron Activation (PFTNA) technology.
Monitor raw material feed to crushers, mills, screens, preparation plants, and coal-fired power plants to help ensure precise feeding of process materials and maintain product quality. Thermo Scientific Ramsey conveyor belt scales provide vital information for the effective management and efficient operation of cement raw material handling.
Reduce material waste, help maintain blend consistency, increase profits, and meet customer specifications. Thermo Scientific weighbelt feeders are used in cement plants to provide a consistent feed of raw materials to maintain their desired proportions in the raw mix, which helps reduce material waste and help maintain blend consistency.
Protect expensive crushers, conveyors and other process equipment from damage by tramp metal. Thermo Scientific Ramsey Oretronic IV Tramp Metal Detectors detect all types of metallic scrap that conventional metal detectors may miss.
Get on-the-spot elemental analysis of quarry samples. Thermo Scientific Niton handheld XRF analyzers can also determine whether the refractory anchors are of the required grade of steel. Analysis can be carried out at the time of material receipt to reduce the chance of material mismatches during procurement.
Learn everything you need to know to improve product quality, boost efficiency, lower productions costs, and create safer, cleaner cement production lines. Download the free eBook, A Practical Guide to Improving Cement Manufacturing Processes and Production.
Belt conveyer scales are an important part of a mining operation’s bulk material handling system. A belt scale system monitors production output and inventory, or regulates product loadout, while providing vital information for the effective management and efficient operation of your business. There are many options to consider when selecting a belt scale system. How do you know which one is right for your application?
Check out the Belt Scale Selection Guide
Related products used in mining cement raw materials
Conveyor protection switches for bulk material handling equipment are important tools in helping to prevent accidents, protect equipment, and reduce unscheduled shutdowns. These industrial process monitoring switches are used for position information, control signals, and to identify potentially hazardous situations with your process equipment.
Monitor solid and liquid flows in a variety of settings and environments. We offer continuous level measurement devices that penetrate dust, steam, and vapors to provide precise, reliable readings. Our level sensors prevent bin overflows, plugged chutes, and empty bins, and our microwave or nuclear point level detection and control switches monitor the silo or bin level above the weighfeeder.
Our online elemental analyzers utilize a Neutron source and also generate Gamma rays as part of the analytical process. We offer radiation detection pagers to assess the possible radiation exposure and ensure that these stay within acceptable limits.