Reduce hands-on time and simplify laboratory testing during your analytical process with the Thermo Scientific Gallery Plus Discrete Analyzer. This integrated platform provides two measurement techniques for photometric (colorimetric and enzymatic) and electrochemical (pH and conductivity) analysis, which can be run simultaneously. Parallel determination of multiple analytes from a single sample as well as the presence of several automated features ensures analytical efficiency. The Gallery Plus discrete analyzer can accommodate 108 samples and 42 reagents in separate sample and reagent disks, with the capability to run up to 350 tests/hour.
Key features
Unique disposable Decacell
- The low cost, disposable Decacell cuvettes used are 10 independent reaction cells mounted together for true discrete analysis
- No need for washing- Improves throughput
- No carry over – improves result reliability
- Reduced sample and reagent consumption: 2–240 μL
- Lowest waste generation and disposal cost
- Reduced cost per analysis
High throughput analyzer
- True direct read discrete analyzer
- Increased throughput with a sample capacity of up to 350 tests/hour
- Optional ECM unit for parallel pH and conductivity measurement
- 12 filter positions - covering wide spectral range from 340 to 880 nm
- Up to 20 different chemical parameters per sample
- Measurements cover a large concentration range with excellent reproducibility - Sensitivity to the ppb level
Versatile and ease of use
- Pre-defined ready to use methods
- More than 50 ready to use enzymatic and colorimetric reagents
- Automated calibration from single stock standard
- Automatic dilution of over range samples
- Wide incubation temperature range from 25 °C to 60 °C
- User-specific applications can be developed to meet individual testing needs
User interface and data management
- Workflow based operation— suitable for all users’ levels
- LIMS connectivity
- User administration
- Traceability - Built-in barcode readers for samples and reagents
- Real-time QC program with multiple, user-definable Westgard rules
- QC charts - Out-of-specification control results flagged
- Instrument validation using IQ Kit
Low cost of ownership
- Xenon source lamp – Savings over lifetime of the instrument
- Ready to use Gallery system reagents eliminate time-consuming reagent preparation, freeing operators for other tasks
- Unique low volume cuvette design for less reagent waste and potentially lowering operation costs
- Contamination-free analysis using disposable cuvettes
- Accommodates a variety of sample cups and primary tubes
Benefits
Anyone can perform the required wet chemical analyses regardless of their chemistry background. Gallery discrete analyzers are easy to use and maintain, the workflow-based operation is suitable for all users’ levels.
Reduce the risk of rerunning samples due to carry over effect with true discrete measurement and disposable cuvettes. Thermo Scientific system reagents are ready to go, saving the technician’s time and reduces preparation errors, ensuring confidence in the quality of results.
Thermo Scientific Gallery and Gallery Plus discrete analyzers are easy to use, automated systems that allow laboratories to simplify their testing with the dual benefits of time and cost savings. Because the instruments operate without manual intervention, you can enjoy walkaway productivity and focus on more strategic tasks.
The unique low volume cuvette design guarantees smallest reagent consumption, 2-240 µL, minimizes reagent waste, and as a result reduces reagent costs. Optimized reagent kit sizes and on-board stability further minimize the amount of waste produced and increase cost efficiency.
Fully automated, rapid, random access multiparameter analyzer can perform up to 350 photometric tests per hour and up to 67, parallel, pH and conductivity measurements with optional ECM unit. Parallel determination of several analytes from a single sample as well as the presence of several automated features ensures analytical efficiency.
Anyone can perform the required wet chemical analyses regardless of their chemistry background. Gallery discrete analyzers are easy to use and maintain, the workflow-based operation is suitable for all users’ levels.
Reduce the risk of rerunning samples due to carry over effect with true discrete measurement and disposable cuvettes. Thermo Scientific system reagents are ready to go, saving the technician’s time and reduces preparation errors, ensuring confidence in the quality of results.
Thermo Scientific Gallery and Gallery Plus discrete analyzers are easy to use, automated systems that allow laboratories to simplify their testing with the dual benefits of time and cost savings. Because the instruments operate without manual intervention, you can enjoy walkaway productivity and focus on more strategic tasks.
The unique low volume cuvette design guarantees smallest reagent consumption, 2-240 µL, minimizes reagent waste, and as a result reduces reagent costs. Optimized reagent kit sizes and on-board stability further minimize the amount of waste produced and increase cost efficiency.
Fully automated, rapid, random access multiparameter analyzer can perform up to 350 photometric tests per hour and up to 67, parallel, pH and conductivity measurements with optional ECM unit. Parallel determination of several analytes from a single sample as well as the presence of several automated features ensures analytical efficiency.
Specifications
Thermo Scientific Gallery discrete analyzers are fully automated, sample oriented random-access analyzers. Many different tests can be run simultaneously from a single sample. Panels of system applications are predefined into the analyzer, e.g., for food, beverage, enzymes, water, environmental and bioprocess quality control and testing. However, new user-specific applications can be defined. Gallery discrete analyzers employs colorimetric end-point and kinetic, as well as turbidimetric and bichromatic reactions with or without sample blanking. System supports automated standard addition measurement method for complex sample matrices.
- Continuous access to samples, reagents, and cuvettes without interruption of routine workflow
- Automatic identification via internal barcode reader
- Clearly displayed real-time reagent volume and remaining test capacity. Up to four reagent additions per test
- Dilutions and repeat analyses are handled automatically
- Measurements cover a large concentration range with excellent reproducibility
- Ability to achieve detection of very low levels (ppb)
- Many standard and USEPA approved methods for waste water and drinking water
Ordering table
Gallery Plus Discrete Analyzer | Gallery Plus Discrete Analyzer | ||
|---|---|---|---|
| Request quote | Request quote | ||
| Discrete analyzer | Measurement method | Rapid, photometric analysis of multiple analytes for various applications | Rapid, photometric and electrochemical analysis of multiple analytes for various applications |
| Cuvettes | Discrete disposable cuvettes | ||
| Performance | Cuvettes capacity | On board capacity 360 measurement cells | |
| Cuvette cleaning | Disposable cuvettes, saves time, improves throughput, precision and accuracy | ||
| Cross contamination | Nil | ||
| Sample capacity | 108 samples | ||
| Reagent capacity | 42 positions | ||
| Reagents per test | Up to 4 reagents addition | ||
| Tests/hour | 350 tests/hour | ||
| Parameters per sample | Up to 20 parameters | ||
| Samples & reagents | Sample volume | 2–120 μL (possibility to extend up to 240 μL) | |
| Reagent volume | 2–240 μL | ||
| Sample containers | 0.5, 2.0 and 4.0 mL sample cups 5.0, 7.0 or 10.0 mL sample tubes | ||
| Technical information | pH range | 2–12 | |
| Conductivity range | 20 μS/cm–112 mS/cm | ||
| Wavelength range | 340 nm–880 nm | ||
| Filter disc | 12 position | ||
| Linear range | 0 to 3.5A | ||
| Resolution | 0.001A | ||
| Source lamp | Long-life Xenon lamp | ||
| Bitterness test | — | ||
| Measurement temperature | Adjustable between 25 °C and 60 °C; Default setting at 37 °C | ||
| Auto analysis | Automated calibration | ✓ | |
| Auto dilution of over range samples | ✓ | ||
| Barcode reader for samples and reagents | ✓ | ||
| Auto start up & shutdown | ✓ | ||
| Urgent samples | Continuous access to samples, reagents, and cuvettes without interrupting the test cycle | ||
| User interface | Bi-directional LIMS | ✓ | |
| Calibration | Factor, bias, linear, logit-log, spline, 2nd order and point-to-point | ||
| QC check | Real-time QC program with multiple and variable rules; Control frequency user-definable | ||
| Out of limit | Results flagged | ||
| Hardware | Dimension | 94 cm W × 70 cm D × 62 cm H (closed) 94 cm W × 70 cm D × 130 cm H (open) | |
| Weight | 110 kg (242 lbs) | ||
| Part Number* | MG9862 0001 | MG9862 2001 | |
Discrete analyzer market applications
Mastering Beer Analysis
For brewers, the compounds of interest range—from inorganic ions, organic acids and hop bittering principles that contribute to overall taste and bitterness—to proteins, carbohydrates and alcohols that are monitored to determine the extent of fermentation. Dedicated for beer, malt, and wort analysis, the Gallery Plus Beermaster discrete analyzer automates labor-intensive and time-consuming tests.
Simplified Wine Analysis
Wineries needs a reliable analytical instrumentation for multi-parameter analysis, that enables lab personal without technical or chemistry knowledge to carry out the routine wine analysis. Thermo Scientific Gallery discrete analyzers together with ready to use system reagents makes the overall wine analysis simple, accurate and reliable. The ready to use wine reagents are specifically developed for rapid and cost-effective analysis of juice and wine.
High throughput Environmental Analysis
Drinking water safety is clearly a critical priority. The U.S. Environmental Protection Agency, through the Safe Drinking Water Act, has established national health based standards that address both man-made and naturally occurring contamination. Labs must ensure municipal or industrial wastewater is safe before its discharge into surface waters in accordance with regulations in the U.S. EPA Clean Water Act (CWA). Compliance requires consistent, accurate measurement of water parameters. Our discrete analyzers automate photometric and electrochemical analysis providing fast, reproducible results that allow laboratories to measure multiple analytes simultaneously.
Industrial water analysis
To eliminate corrosion and scaling that can damage valuable industrial components and to ensure regulatory compliance, reliable water analysis must be performed continuously.
Resources
Solutions to resolve analytical challenges in industrial and waste water processing infographic
Reliable water analysis is critical to eliminating corrosion and scaling that can damage valuable industrial components and to ensure regulatory compliance. Download the infographic to learn about our solutions to the top three analytical challenges in industrial water and waste water processing.
Solutions for Carbohydrate Analysis Webinar Series
Robust and Reliable, an Automated Analyzer Increases Efficiency at a Napa Valley Winery
Merryvale Family of Wines selected a robust, reliable, repairable instrument in order to automate their testing procedures. As a result, Starmont winery reduced the amount of time consumed for testing and gained the ability to attend to other vital tasks, especially during the busy harvest season.
No records were found matching your criteria