Thermo Scientific crimp top vials for HPLC and GC are available in 8mm, 11mm, or 13mm crimp border diameter in clear and amber glass and plastic. Microsampling options for small-sample-volume injections include one-piece microvials and standard inserts added to crimp top vials. Crimp vials are sealed with aluminum caps with septa using crimper and decrimper tools (manual or electronic). The vial selected depends on your sample size and instrument compatibility (see autosampler compatibility chart).
Snap vials sealed with snap caps are easy to apply and remove without the need for crimping or de-capping tools. These chemically inert polyethylene caps are an economical choice for most routine HPLC applications but have low sealing property. Snap caps have a large open area allowing easy needle penetration across the entire vial opening. Aluminum crimp caps can also be used to seal snap-it vials (using a crimper).
Thermo Scientific crimp top & snap cap vials and closures
8mm Crimp Vials | 8mm Crimp Caps, Septa, Inserts |
---|---|
Instrument compatibility
- Agilent
- Beckman
- Carlo Erba
- CTC
- Fisons
- PerkinElmer
- Shimadzu
- Thermo Scientific
- VWR (Merck)/Hitachi
11mm Crimp/Snap Vials | 11mm Crimp/Snap Caps, Septa & Inserts | 11mm Crimp/Snap Vial Kits |
---|---|---|
11mm Wide Opening Crimp/Snap Top Vials
- Superior quality 1st hydrolytic class glass (Type 1) in clear and amber meets all requirements of Pharm. US, EU, JPN
- Aluminum crimp seals with prefitted septa for all 11mm crimp top and snap cap vials
- Pre-assembled caps and septa are convenient and minimize contamination from handling
- Aluminum seals must be applied with a crimping tool
- Microvials and inserts are available for microsampling options
Instrument compatibility
- Most HPLC and GC autosamplers
4mL, 15x45mm crimp/snap cap vials and inserts
- Superior quality 1st hydrolytic class glass (Type 1) with a 13mm crimp/snap-ring finish
- Use with 13mm aluminum Seals or Kim-Snap closures
- Polyspring inserts are self-aligning and provide a cushion against needle contact
National 13mm Crimp/Snap Caps and Septa
- Pre-assembled caps and septa are convenient and minimize contamination from handling
- Aluminum crimp closures provide a secure leak-resistant seal
- Aluminum seals must be applied with a crimping tool
- Synthetic PTFE/red rubber seal is specially formulated for improved background performance
- Kim-Snap closures provide a tight seal without the need of a crimper
Recommended for the following instruments:
- Thermo Scientific
- Shimadzu
- Spark Holland
- Varian
- VWR (Merck)/Hitachi
- Waters (Wisp 48 Position Carousel)
Consider this screw top vial as a crimp top alternative
Consider Thermo Scientific 2 mL SureStop Screw Top Vials with AVCS Caps as a secure alternative to crimp top vials. These specially designed screw top vials create an optimal seal with performance data similar to traditional crimp vials. See this technical note for performance data. Screw top vials are convenient and eliminate the need for crimping and decrimping tools.