Search Thermo Fisher Scientific
Maximize your sample performance and have fewer failed assays with the Thermo Scientific Chromacol GOLD Grade Vials. These vials offer the most state-of-the-art technology in the industry for the analysis of critical analytes during low detection limit assays, and when sample quality, reproducibility, security, and integrity are of upmost importance.
Increasingly sensitive chromatography and mass spectrometry systems demand reliable vials which can routinely deliver the ultimate in purity and the lowest surface activity possible. Sample security, and confidence in data integrity, are paramount in trace analysis and can only be achieved with the highest glass vial quality.
For this reason, we have introduced a higher grade of glass with the lowest levels of alkaline materials. You can maximize your analytical performance and have fewer failed assays with the Chromacol GOLD-Grade vials. These premium vials are manufactured in exacting conditions and at optimum surface-saving temperatures to ensure the maximum degree of inertness within the low-alkaline glass.
Increase productivity and save time.
Achieve confidence in results.
Reduce cost of analysis.
Inert Vial/Insert Description | Product Number |
---|---|
8mm Crip-Top Vials | |
0.1mL Crimp Top Round Bottom Vial – Clear, GOLD-Grade | 01-CVG |
0.2mL Crimp Top Tapered Vial – Clear, GOLD-Grade | 02-CTVG |
0.3mL Crimp Top Round Bottom Vial – Clear, GOLD-Grade | 03-CVG |
2mL, 12x32 Standard Opening Screw-Thread Vials | |
2mL Screw Top Vial – Clear, GOLD-Grade | 2-SVG |
1.1mL Screw Top Tapered Vial – Clear, GOLD-Grade | 1.1-STVG |
9mm Wide Opening, Screw-Thread Vials and Insert | |
2mL Wide Mouth Screw SureStop GOLD-Grade Vial with Write-on Patch - Clear | 2-SVWGK |
0.2mL Screw Top Fixed Insert Vial – GOLD-Grade with Write-on Patch ( Micro+) | 02-FISVG |
200µL Insert, GOLD-Grade Glass | 02-MTVWG |
2mL, 12 x 32, 11mm Crimp-Top Vials | |
2mL Crimp Top Vial – Clear, GOLD-Grade | 2-CVG |
1.1mL Crimp Top Tapered Vial - Clear | 1.1-CTVG |
0.2mL Crimp Top Fixed Insert Vial – GOLD-Grade with Write-on Patch ( Micro+) | 02-FIVG |
200µL Insert, GOLD-Grade Glass Insert | 02-MTVWG |
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
Select Chromatography Consumables and Columns for your Pharma, Biopharma, Clinical, Food & Beverage or Environmental workflow.