Essential IC that doesn’t compromise
Our chemists and engineers have been manufacturing high-quality ion chromatography systems since 1975. Since we have been doing ion chromatography the longest, no one else has more experience for you to leverage. Whether you perform standard water analysis applications or need an instrument for the academic setting, we have the right instrument for your lab. Speak to an expert to find the system that’s right for you or order online for faster delivery.
![]() Dionex Easion IC System | ![]() Dionex Aquion IC System | Why it matters | |
---|---|---|---|
Buy now Request Quote | Buy now Request Quote | ||
Description | Thermo Scientific Dionex Easion IC system brings simplicity to everyday analyses by providing everything you need in one small package. No additional pumps or ancillary items are required to operate Easion. Anion and cation chemistry kits take the guess work out of setup and running. | Thermo Scientific Dionex Aquion IC system brings a new level of robustness and reproducibility to common ion chromatography analyses. Upgradeability allows for optional features to be added at anytime, so it is flexible if your analytical needs or methods change. | |
Dimensions | H 13” W 9.5” D 15.7” | H 22.1” W 8.8” D 21” | Saves you space in your lab so you can easily accommodate other technologies. |
Software | Thermo Scientific Chromeleon Version 7.2.10 | Thermo Scientific Chromeleon 7.2 and 7.3 | Aquion is compatible with the newest version of Chromeleon |
Flow Path Material | PEEK | PEEK | Inert material compatible with IC eluents |
Suppression | Only Chemical Suppression | Both (Chemical & Electrolytic) | Electrolytic suppressors require no regenerant, meaning there are fewer reagents to make, no acid exposure, and more consistent results |
Column Heater | No | Yes, optional | Heater gives increased precision and enables applications that run at elevated temperatures (including some basic EPA methods) |
Eluent Degassing | No | Yes, optional | Degasser reduces baseline noise |
Auxiliary Valve | No | Yes, optional | Enables automated sample preparation |
Leak Detection | No | Yes, built in optical sensor | Helps with troubleshooting |
Autosampler-compatible | Yes (AS-DV or AS-AP) | Yes (AS-DV or AS-AP) | Enables walkaway operation when analyzing multiple samples |
CMD SchemaApp code