Learn what's new in the latest release of Amira-Avizo Software
Thermo Scientific Amira-Avizo Software Version 2019.3 rolls out exciting new tools and enhancements. Read on for a quick introduction to these features and links to resources offering more information.
We keep enriching Amira-Avizo Software’s unsurpassed processing toolbox with new Artificial Intelligence-based processing tools
While advanced Texture Supervised Classification tools and Color Auto Classification, powered by Machine Learning, were released in Amira-Avizo Software version 2019.2 and PerGeos Software version 2019.2, Amira-Avizo Software version 2019.3 provides a powerful deep learning environment.
The use of DL-based tools in Amira-Avizo Software is a major leap forward and enriches the processing capabilities by allowing the mixing of both traditional and AI-based algorithms.
Resource datasets and examples of trained models are available for download from the Xtra library website.
Data courtesy of Cardona A, et al. 2010. An Integrated Micro- and Macro-architectural Analysis of the Drosophila Brain by Computer-Assisted Serial Section Electron Microscopy. PLoS Biol 8(10): e1000502. doi:10.1371/journal.pbio.1000502.
With the 2019.3 release, a new online resource portal is available, providing a collection of add-ons (recipes, scripts, demos) dedicated to specific use cases and workflows, that will help you improve your daily use of Amira-Avizo Software and dramatically increase your efficiency.
The Xtra Library will be populated with new add-ons continuously, independent of the release cycle of the products.
This new filter enables enhancement of structures such as balls (dots, bubbles, spherical cells), rods (filaments, fibers), and planes (membranes, cracks), relying on either the eigenvalue analysis of the Hessian or the Structure Tensor.
The filter can operate at a range of scales and is thus able to highlight structures of different sizes.
Read the full Release Notes
The Release Notes allow you to gather a deeper knowledge about this new version of Amira-Avizo Software. They include more technical explanations, compatibility notes, operating systems Amira-Avizo Software runs on and a list of solved issues.
Amira-Avizo Software Upgrades
Amira Software, Avizo Software and Avizo Software for Industrial Inspection 2019.3 are regular upgrades and will be provided free-of-charge to all users under valid maintenance.
The Structure Enhancement Filter module is available under the Avizo license and Amira XImagePAQ extension license.
These new versions also bring many improvements and fixes to known issues.
What was new
Amira-Avizo Software 2019.2
Released on June 2019
Amira-Avizo Software 2019.1
Released on March 2019
Amira Software 6.7
Avizo Software 9.7
Released on October 2018