Search Thermo Fisher Scientific
Thermo Scientific Amira-Avizo Software Version 2019.4 rolls out exciting new tools and enhancements. Read on for a quick introduction to these features and links to resources offering more information.
2 new color maps are introduced in this version, invertedGrayScale and volrenYellowInverted. Your 2D and 3D visualization will benefit from these new colormaps when the background information in the image has a higher intensity than the structures of interest.
The module Convert Image Type has been enhanced to offer a simplified interface for normalizing intensities before casting, using explicit ranges and appropriate default values.
Absolute script path evolved to relative path in order to avoid the previous path update needed.
What is a relative path: A relative path starts from some given working directory, avoiding the need to provide the full absolute path.
With this version, when saving the recipe, the absolute path is replaced by a relative one if the change is possible. To do so, the recipe should be saved next to or below the script in the file tree. Of course, the file tree should be kept as is when sharing.
The 2019.4 version of Amira-Avizo for EM Sytems now supports the FEI TIFF color images.
With this new 2019.4 version, you can make statistical analysis for column containing scalar values using the new method ‘HxSpreadSheetInterface.Column.asarray’
You can now send and receive Python commands via TCP/IP sockets from a Client to a Server that will interpret the command. It allows you to create Python script which generates a complete compute module workflow and execute it on the server side.
The albumentations is now standard in the Python environment created with Deep Learning packages.
The Release Notes allow you to gather a deeper knowledge about this new version of Amira-Avizo Software. They include more technical explanations, compatibility notes, operating systems Amira-Avizo Software runs on and a list of solved issues.
Amira Software, Avizo Software, Avizo Software for Industrial Inspection and Amira-Avizo Software for EM Systems 2019.4 are regular upgrades and will be provided free-of-charge to all users under valid maintenance.
These new versions also bring many improvements and fixes to known issues.
Amira-Avizo Software 2019.3
Released on November 2019
Amira-Avizo Software 2019.2
Released on June 2019
Amira-Avizo Software 2019.1
Released on March 2019