Automated STR-based human identification
Product description | Part number category | Part number | Price |
RapidHIT System Instrument | Instrument | 100010RH | Request a Quote |
RapidHIT GlobalFiler Express 3-Sample Cartridge Kit (12 Samples per Kit) | Consumables | 400084RH | Request a Quote |
RapidHIT GlobalFiler Express 7-Sample Cartridge Kit (28 Samples per Kit) | Consumables | 400086RH | Request a Quote |
RapidHIT GlobalFiler Express 8-Sample Cartridge Kit(32 Samples pr Kit) | Consumables | 400106RH | Request a Quote |
RapidHIT NGM SElect Express 7-Sample Cartridge Kit(28 Samples per Kit) | Consumables | 400090RH | Request a Quote |
RapidHIT NGM SElect Express 3-Sample Cartridge Kit Plus (12 Samples per Kit) | Consumables | 400102RH | Request a Quote |
RapidHIT NGM SElect Express 8-Sample Cartridge Kit(32 Samples per Kit) | Consumables | 400105RH | Request a Quote |
RapidHIT, Storage Cartridge Kit (10 uses) | Consumables | 400061RH | Request a Quote |
RapidHIT Blank Cartridge Kit (4 pack; 20 runs per cartridge) | Consumables | 400087RH | Request a Quote |
Description:
The IntegenX by Thermo Fisher Scientific RapidHIT System is the first fully-automated, sample-to-answer system for STR-based human identification. RapidHIT is available to forensic scientists in booking stations, laboratories, and in the field home or abroad.
The self-contained human identification system produces standardized DNA profiles from buccal swabs and other human samples in under two hours. Requiring only three minutes of hands-on time to generate up to eight DNA profiles in one run, the easy-to-use system fits into any forensic workflow. The RapidHIT system uses NDIS-approved Applied Biosystems GlobalFiler Express chemistry, and the output from the system is compatible with local, state and national human identification databases.
- >90% full profile and CODIS success rate
- 1 to 8 samples in one run
- Process any swab type and certain crime scene samples directly in sample cartridge
- Requires only 3 minutes of hands-on time, less than 2 hours time to results
Specifications:
Dimensions and weight | 73 x 71 x 48 cm (28.5 x 28 x 18 in) 81.5 kg (180 lbs) |
Performance | Up to 8 samples per run Processing time under 2 hours Allelic ladder control Applied Biosystems GlobalFiler Express–24 markers including expanded core CODIS loci Applied Biosystems AmpFLSTR NGM SElect Express–17 markers |
Certificates | Manufactured under ISO 9001 CE and TUV |
Operating environment | 18–30ºC (59–86ºF) 15–75% relative humidity Elevation up to 2000 m (6500 ft) 100–240 VAC, 50/60 Hz, 120V, 400W (peak at 600W), recommended 1000 VA uninterruptible power supply |
Resources
Product literature
Publications
- Investigative leads from DNA: Casework experience from the IntegenX RapidHIT 200 System
- Generating Rapid DNA Profiles from Crime Scene Samples Commonly Encountered in the United Arab Emirates
- Evaluation of the RapidHIT 200 System: A comparative study of its performance with Maxwell DNA IQ/Identifiler Plus/ABI 3500xL workflow
- Evaluation of the RapidHIT 200, an automated human identification system for STR analysis of single source samples
- Developmental validation of a fully integrated sample-to-profile rapid human identification system for processing single-source reference buccal samples
- An evaluation of the RapidHIT system for reliably genotyping reference samples
- Developmental validation of the GlobalFiler express kit, a 24-marker STR assay, on the RapidHIT System
- DNA investigative lead development from blood and saliva samples in less than two hours using the RapidHIT Human DNA Identification System
- Performance of the RapidHIT 200
- RapidHIT 200, a promising system for rapid DNA analysis