Product description | Part number category | Part number | Price |
RapidHIT ID ACE GlobalFiler Express 150 Sample Cartridge Kit | Consumables | 400093RH | Request a Quote |
RapidHIT ID ACE NGM SElect Express 150 Sample Cartridge Kit | Consumables | 400095RH | Request a Quote |
RapidHIT ID EXT GlobalFiler Express 150 Sample Cartridge Kit | Consumables | 400127RH | Request a Quote |
RapidHIT ID Primary Cartridge 250 | Consumables | 400118RH | Request a Quote |
RapidHIT ID ACE GlobalFiler Express 125 Sample Cartridge Kit | Consumables | 400121RH | Request a Quote |
RapidHIT ID ACE NGM SElect Express 125 Sample Cartridge Kit | Consumables | 400122 | Request a Quote |
RapidHIT ID EXT GlobalFiler Express 125 Sample Cartridge Kit | Consumables | 400115RH | Request a Quote |
RapidHIT ID EXT NGM SElect Express 125 Sample Cartridge Kit | Consumables | 400117RH | Request a Quote |
Description:
The IntegenX by Thermo Fisher Scientific RapidHIT ID consumables support legacy IntegenX systems.
The compact, easy-to-use IntegenX RapidHIT ID System generates lab-quality forensic DNA profiles in virtually any setting in less than 90 minutes. Processing a buccal swab is simple, requiring less than one minute of hands-on time. DNA profiling has never been this fast or this easy.
RapidHIT ID generates DNA STR profiles using NDIS-approved chemistry, developed to meet FBI Quality Assurance Standards (QAS) and criteria for Rapid DNA analysis. RapidHIT ID data formats are compatible with established DNA databases (e.g., CODIS, NDNAD, NCIDD).
- Compact instrument footprint
- Lab-quality DNA profiles in <90 minutes
- Less than 1 minute of hands-on time
Specifications:
Performance | Processing time >90 minutes Applied Biosystems GlobalFiler Express–24 markers, including expanded core CODIS loci Applied Biosystems AmpFLSTR NGM SElect Express–17markers |
Operating environment | 15–35ºC (59–95ºF) 20–90% relative humidity Altitude up to 3,000 m (9,842 ft) 100–240 VAC, 50/60 Hz, 120V, 600W |
Resources
Publications
- Evaluation of a rapid DNA process with the RapidHIT ID system using a specialized cartridge for extracted and quantified human DNA
- Internal validation of the RapidHIT ID system
- Validation of a rapid DNA process with the RapidHIT ID system using GlobalFiler® Express chemistry, a platform optimized for decentralized testing environments
Materials safety data sheets
For Forensic or Paternity Use Only.