Meet quality standards with versatile elemental analysis
Perform rapid, accurate analysis of trace and alloying elements throughout production. Our OES metal analyzers can be used at different production stages, while XRF is used for analysis of raw materials, slags and alloys. Analysis quality and speed can be further enhanced with our robotics-based platform that fully automates sample preparation, sample transfer and spectrometer operation. Our SEM/EDX system provides fast information for the inclusion size, shape, image, and composition a variety of product samples.
Obtain precise spectrochemical analysis of metals for quality control and production. Demanding steel plants, pure metals producers and companies needing the best determination of all the elements turn to the Thermo Scientific ARL iSpark Series Optical Emission Spectrometer for high accuracy analysis of iron and steel from trace to alloying element levels. Optional methods using the Spark-DAT algorithms allow ultra-fast analysis of microinclusions, such as Al2 O3, CaO, Al2 O3 -CaO, MnS and CaS in steel.
Perform fast visual troubleshooting with the Thermo Scientific ARL easySpark Metal Analyzer. Designed for for small to mid-size foundries and metals processing companies, or as back-up instrument, this bench top OES analyzer offers simple operation with with wide elemental coverage from trace to percent level.
Save money and achieve quality control in your industrial process with full simultaneous and sequential capabilities integrated into one instrument.
- Analyze Be to Am in solids, fused beads or pressed powders.
- Unbeatable speed, precision and light-element sensitivity.
- Configurable for your applications with choice of goniometers, monochromators and XRD system.
- Choice of generator power depending on the analysis speed required.
- Safe and reliable loading of samples with X-ray tube above the sample.
Drive process control laboratory efficiencies in iron and steel applications. The Thermo Scientific ARL SMS-3500 Automation for Twin OES or XRF offers a complete laboratory workflow automation solution. Double sample preparation machine support reduces response times, increases sample processing cadence and improves the availability of automatic sample preparation in highly critical production control environments.
Monitor the quality of incoming material and finished product with improved inclusion analysis and process control. The Explorer 4 Analyzer is the fourth generation of dedicated SEM/EDX solutions for industrial environments and data acquisition and features features a significant improvement in source life and ability to size smaller inclusions and particles for today's manufacturing environment. Report to ASTM E45, ASTM 2142, JIS G 0555, ISO 4967, DIN 50 602, and EN 10247 inclusion ratings for steel.
Video spotlight: analytical instruments used in steel manufacturing
Explorer 4 process map
Download our free eBook to learn about Thermo Scientific products that improve each stage of the steel manufacturing production process to help you manufacture clean steel that meets the most stringent customer requirements.