A whole hospital approach to radiation protection
Radiation monitoring is an integral part of good health physics practice at every medical facility performing any radiation therapy activities. It is the core discipline of RSOs, Health Physicists, Medical Physicists, Radiologists, and Nuclear Medicine Techs to be acutely aware of radiation exposure rates for staff and patients.
Thermo Fisher Scientific offers a wide range of contamination survey meters, area monitors, and dosimeters for Radiation Safety Officers to keep track of critical Radiation Controlled Areas (RCA) that require radiation monitoring.
Radiation monitoring solution videos
- The Thermo Scientific FHT 6020 Area Monitor Display and Alarms monitors the radiation level of a room in areas such as cancer treatment centers where radiation therapy is part of the daily work routine.
- Measures and assesses the level of gamma and neutron radiation; features audible and visible alarms in case of emergency.
- Optional remote light tower alerts from a distance if radiation is detected.
- The Thermo Scientific FHT 762 Wendi-2 Wide Energy Neutron Detector delivers high sensitivity and an excellent energy and angular response.
- Fits to FHT 6020 area monitor and FH 40 G survey meter.
- The Thermo Scientific FH 40 G Multi-Purpose Digital Survey Meter is highly configurable with external detectors to support a wide range of applications.
- Optional plug and play probes available for neutron measurement, alpha and beta contamination readings, and artificial gamma radiation detection in variable natural radiation fields.
- The Thermo Scientific ViewPoint Enterprise Remote Monitoring System is a comprehensive software solution integrating personnel dosimetry and facility monitoring for a complete, realtime picture of facility operations.
- Simplified instrument data management and complete archival simplifying analysis, enabling informed decisions and report creation.
- Configurable with a variety of instruments.
- Thermo Scientific Dosimetry Services offer you accurate measurement of radiation dose exposure in the workplace, ensuring personnel safety in any environment where radiation exposure is an issue.
- We send you passive dosimeter badges, your employees wear the badges for a period of time and then the badges are shipped back to our facility for analysis and online reporting.
- Adding RadSight Live delivers live dose rate monitoring of a team. Data can be viewed from any computer on the network.
- The Thermo Scientific EPD TruDose Electronic Personal Dosimeter delivers ultra-precise, real-time dose reading and reporting.
- Multidetector technology measures both gamma and beta radiation.
- Improved dose rate range and unprecedented sensitivity, providing you peace of mind in the accuracy of exposure.
- Thermo Scientific RadEye B20 and B20-ER Multi-Purpose Survey Meters are simple, robust, reliable contamination and gamma dose rate measurement tools for characterizing alpha, beta, gamma and X-ray radiation.
- Excellent for nuclear medicine and incoming/outgoing package inspection.
- The Thermo Scientific RadEye SX Survey Meter for Scintillation Detectors is a multi-purpose meter for external scintillator counter tubes.
- General count rate and surface contamination measurements as well as dose rate measurements.
- The Thermo Scientific IPM96 Whole Body Contamination Monitor and the Thermo Scientific iPCM12 Installed Personnel Contamination Monitor both can be installed at area perimeters to help prevent the spread of radioactive contamination throughout your facility.
- Designed to detect both alpha and beta radiation.
- Can be deployed at hospitals, PET facilities, radiological centers where patients are injected with radioactive isotopes, or radiopharmaceutical departments.
- Thermo Scientific RadHalo RDP and FM Spectroscopic Area Monitors offer quick, real-time monitoring and measurement of gamma radiation.
- Highly configurable.
- Conveniently managed with monitoring software or on your mobile device.
- The Thermo Scientific TPM-903C Transportable Radiation Portal Monitor provides quick, accurate head-to-toe screening in emergency response scenarios.
- Used to monitor personnel entering and existing Radiation Control areas or in case of an emergency radiological event.
- The Thermo Scientific RadEye SPRD-ER Personal Radiation Detector sets a new standard in the detection of hidden or illicit radiation through fine-tuned detector sensitivity and smarter alarming.
- Can help staff more closely identify radioactive isotopes in medical waste.
- Thermo Scientific RadHalo RDP and FM Spectroscopic Area Monitors can be rapidly deployed in response to a nuclear accident.
- Monitor and manage with your mobile device with flexible monitoring software.
- Engineered for the harshest environments.
- The Thermo Scientific ViewPoint Enterprise Remote Monitoring System centrally processes and analyzes data from radiation, environmental and general purpose detectors to give a complete picture of your environment.
- Comprehensive instrument integration (Thermo Scientific and third-party instruments)
- The Thermo Scientific FHT 6020 Area Monitor Display and Alarms monitors the radiation level of a room in areas such as cancer treatment centers where radiation therapy is part of the daily work routine.
- Measures and assesses the level of gamma and neutron radiation; features audible and visible alarms in case of emergency.
- Optional remote light tower alerts from a distance if radiation is detected.
- The Thermo Scientific FHT 762 Wendi-2 Wide Energy Neutron Detector delivers high sensitivity and an excellent energy and angular response.
- Fits to FHT 6020 area monitor and FH 40 G survey meter.
- The Thermo Scientific FH 40 G Multi-Purpose Digital Survey Meter is highly configurable with external detectors to support a wide range of applications.
- Optional plug and play probes available for neutron measurement, alpha and beta contamination readings, and artificial gamma radiation detection in variable natural radiation fields.
- The Thermo Scientific ViewPoint Enterprise Remote Monitoring System is a comprehensive software solution integrating personnel dosimetry and facility monitoring for a complete, realtime picture of facility operations.
- Simplified instrument data management and complete archival simplifying analysis, enabling informed decisions and report creation.
- Configurable with a variety of instruments.
- Thermo Scientific Dosimetry Services offer you accurate measurement of radiation dose exposure in the workplace, ensuring personnel safety in any environment where radiation exposure is an issue.
- We send you passive dosimeter badges, your employees wear the badges for a period of time and then the badges are shipped back to our facility for analysis and online reporting.
- Adding RadSight Live delivers live dose rate monitoring of a team. Data can be viewed from any computer on the network.
- The Thermo Scientific EPD TruDose Electronic Personal Dosimeter delivers ultra-precise, real-time dose reading and reporting.
- Multidetector technology measures both gamma and beta radiation.
- Improved dose rate range and unprecedented sensitivity, providing you peace of mind in the accuracy of exposure.
- Thermo Scientific RadEye B20 and B20-ER Multi-Purpose Survey Meters are simple, robust, reliable contamination and gamma dose rate measurement tools for characterizing alpha, beta, gamma and X-ray radiation.
- Excellent for nuclear medicine and incoming/outgoing package inspection.
- The Thermo Scientific RadEye SX Survey Meter for Scintillation Detectors is a multi-purpose meter for external scintillator counter tubes.
- General count rate and surface contamination measurements as well as dose rate measurements.
- The Thermo Scientific IPM96 Whole Body Contamination Monitor and the Thermo Scientific iPCM12 Installed Personnel Contamination Monitor both can be installed at area perimeters to help prevent the spread of radioactive contamination throughout your facility.
- Designed to detect both alpha and beta radiation.
- Can be deployed at hospitals, PET facilities, radiological centers where patients are injected with radioactive isotopes, or radiopharmaceutical departments.
- Thermo Scientific RadHalo RDP and FM Spectroscopic Area Monitors offer quick, real-time monitoring and measurement of gamma radiation.
- Highly configurable.
- Conveniently managed with monitoring software or on your mobile device.
- The Thermo Scientific TPM-903C Transportable Radiation Portal Monitor provides quick, accurate head-to-toe screening in emergency response scenarios.
- Used to monitor personnel entering and existing Radiation Control areas or in case of an emergency radiological event.
- The Thermo Scientific RadEye SPRD-ER Personal Radiation Detector sets a new standard in the detection of hidden or illicit radiation through fine-tuned detector sensitivity and smarter alarming.
- Can help staff more closely identify radioactive isotopes in medical waste.
- Thermo Scientific RadHalo RDP and FM Spectroscopic Area Monitors can be rapidly deployed in response to a nuclear accident.
- Monitor and manage with your mobile device with flexible monitoring software.
- Engineered for the harshest environments.
- The Thermo Scientific ViewPoint Enterprise Remote Monitoring System centrally processes and analyzes data from radiation, environmental and general purpose detectors to give a complete picture of your environment.
- Comprehensive instrument integration (Thermo Scientific and third-party instruments)
Support
We provide a single source for global integrated service, support and parts supply for all your radiation, measurement and security products.
Request service ›