Today’s busy laboratories demand fast answers with high confidence. You can’t afford to slow down or make a mistake, use our FTIR spectrometers to solve your analytical challenges quickly and with maximum confidence. Our instruments simplify laboratory data collection while providing precise, high-quality results on even the smallest samples.
Confidently identify sample components and rapidly verify materials with less effort so you can make critical decisions fast and with less effort. Use the compact Thermo Scientific Nicolet Summit FTIR Spectrometer to quickly see pass/fail QC results on its multi-colored LED LightBar built into the front of the instrument. Reduce the number of steps required to take measurements, automate workflows and generate reports with the Thermo Scientific OMNIC Paradigm Software that guides you through the process.
Combining compact size, affordable price and superior performance, the Thermo Scientific Nicolet iS5 FTIR Spectrometer offers the perfect combination of functionality, fit and value for an entry-level spectrometer. It is the ideal spectrometer for product assurance testing and material identification for industrial, government and academic labs around the world.
Maximize laboratory productivity and quickly collect high-quality spectral data with the Thermo Scientific Nicolet iS20 FTIR Spectrometer. An integrated touch panel allows users of any skill level to quickly analyze challenging samples and a new multi-color LED scan bar gives users immediate feedback on whether the instrument is in idle, collect or alert mode.
Designed to be highly flexible, the Thermo Scientific Nicolet iS50 FTIR Spectrometer can be upgraded from a simple FTIR bench to a fully-automated multi-spectral range system that can acquire spectra from far-infrared to visible. You can initiate novel ATR, Raman and NIR modules at the touch of a button, enabling access to these techniques without manually changing system components.
Characterize new materials with unmatched flexibility and performance using our Thermo Scientific Nicolet iS50R Research FTIR Spectrometers designed for advanced spectroscopic studies. Modulate amplitude and conduct advanced continuous scan, time-resolved step-scan or dual channel spectroscopy applications.
The Thermo Scientific Nicolet iG50 FTIR Spectrometer extends the premium performance of the Thermo Scientific Nicolet iS50 Spectrometer onto a rugged, user-configurable FTIR platform designed for industrial applications. The system is highly configurable, compact, and rack mountable. It offers customized solutions for easy integration into your process monitoring systems, an the ideal choice for OEM integration.
Simultaneously analyze more than 100 gas species using the Thermo Scientific Antaris IGS Gas Analyzer. Developed with input from industry market leaders to solve specific gas analysis needs, this multi-component gas analyzer provides research-grade performance, speed and sensitivity in a package designed specifically for use in plant and process environments.
Bring the power of near infrared to your analytical lab – with minimum hassle and maximum confidence – with the Thermo Scientific Nicolet iS5N FT-NIR spectrometer. Method development is made simple by the powerful Thermo Scientific OMNIC software suite; a familiar environment to thousands of chemists across the globe. Built on the same rugged platform as the Thermo Scientific Nicolet iS5 spectrometer, the Nicolet iS5N spectrometer provides FT-NIR capability in a compact, cost-effective package designed to meet the needs of today’s quality assurance laboratory.