The Thermo Scientific Nicolet iS20 FTIR Spectrometer resets the standard for analytical FTIR instrumentation with its redesigned optical engine and modern industrial design. The Nicolet iS20 Spectrometer provides customers with advanced performance, productivity, and flexibility. Powered by OMNIC software, seamlessly transfer experiment files, custom libraries, TQ methods, accessories and more.
The Thermo Scientific LightDrive optical engine is composed of a rugged and ultra-reliable interferometer, a novel single-point infrared source and a durable solid-state diode laser. These 3 components come with 10-year warranties, setting a new standard for low cost of ownership. The Nicolet iS20 Spectrometer delivers data reproducibility sample after sample with the LightDrive Optical Engine.
Reduce the number of steps it takes to create and run workflows and streamline data analysis with the integrated touch panel designed for fast answers. The multi-colored scan bar helps you know your instrument status at-a-glance. Whether the instrument is ready for analysis, collecting data, or needing attention, you know your next action.
A solid-pulsing green light lets you know your instrument is ready to run.
A moving green light tells you that data is being collected
A solid-pulsing yellow light indicates that your scheduled maintenance date is approaching.
A quick-pulsing red light indicates you should look at your software diagnostics menu for additional information.
The Nicolet iS20 Spectrometer provides the flexibility needed to take your analysis to the next level. When complex problems enter your lab, the Nicolet iS20 is up to the challenge.
- Need to conduct a kinetics analysis for a curing process? Equip your Nicolet iS20 with a high-speed MCT detector.
- Easily measure small particles and defects down to 25 microns in size using the Nicolet iS20 with the Nicolet iN5 FTIR Microscope. Analyze particle compositions with OMNIC Specta multi-component search software.
- The Nicolet iS20 with the TGA-IR module allows you to understand your materials better by correlating infrared and thermal analysis to elucidate composition of mixtures, copolymers and blends.
- The Nicolet iS20 delivers exceptional resolution for gas analysis. We offer a variety of gas cell configurations to meet your sampling needs.
- The Nicolet iS20 Spectrometer is compatible with hundreds of commercially available accessories, expanding your capabilities into the future.
The Nicolet iS20 is powered by Thermo Scientific OMNIC Software. Seamlessly transfer methods, workflows and libraries to the latest version of OMNIC. Also included with the Nicolet iS20 Spectrometer are Thermo Scientific OMNIC Specta for multi-component analysis and OMNIC Anywhere, our cloud-based spectroscopy software, which enables you to analyze and share data anytime, anywhere.
We made compliance easier with automated systems that meet international regulations. Regulated laboratories must have absolute confidence in the answers they provide. Thermo Scientific Security Suite Software allows you to ensure the security and integrity of your data and helps your lab comply with 21 CFR Part 11 or other regulations.
QA/QC
FTIR spectroscopy offers almost unlimited analytical opportunities in many areas of production and quality control determining if a product meets control specifications. An advantage of FTIR is fast data acquisition for both qualitative and quantitative analysis. Create custom methods for each step of QA/QC testing for adherence to strict SOPs.
Pharmaceuticals
Perform raw material identification and deploy methods for product verification with confidence. Qualification tools help you comply with pharmacopeia regulatory requirements. Confidence in instrument installation, operation and performing according to specifications.
Polymer/Chemical
Understand and control chemical properties when designing new materials. Catch changes in additive formulations, degradation of multi-layer polymer composites, weakened packaging materials and effects of process curing times on polymer specifications.
Environmental
Infrared spectroscopy is a valuable technique for monitoring air quality, testing water quality and identifying plastic particles.
The Nicolet iS20 Spectrometer resets the standard for analytical FTIR instrumentation with its redesigned optical engine and modern industrial design. If the 10-year warranties on the interferometer, source and laser are not enough to give you peace of mind, remember Thermo Fisher Scientific has market-leading service and support through Unity Lab Services. We have more than 7,000 service engineers and 1800 technical support specialists worldwide to support you and your lab. Thermo Fisher Scientific is your trusted partner for FTIR spectroscopy solutions.
Tech Direct | Essential & Extended Warranty | Premier | |
| Faster diagnosis and remote resolution to get you back up and running | Decreased downtime with regular maintenance and upkeep | Fastest, priority response time available for critical operations | |
| Specifications | Immediate tech support with on-site response when needed | Comprehensive maintenance with predictable service costs | Instrument availability and uptime prioritization |
| Preventative maintenance with parts and software updates | • | • | • |
| Enhanced Technical Support with priority response and remote resolution | • | • | • |
| On-site response time | 5 days | 3 days | 2 days |
| Corrective maintenance including parts, labor and travel | 10% discount | • | • |
| Service history and site management reviews | • |