Premium fetal bovine serum (FBS)
Fetal bovine serum (FBS) for more stringent applications
Premium FBS offers the least risk of BSE and lower viral risk. These products meet USP/EP guidelines with up to 96 harmonized quality specification tests that include EMA virus testing, USP/EP mycoplasma, endotoxin, performance, biochemical/hormonal profiling, and Oritain™ fingerprinting. Manufactured using triple 0.1 micron filtration.
Product specifications
NEW Premium Plus FBSPremium FBS
Australia
Premium FBS
New Zealand
Premium FBS
United States
Endotoxin≤1 EU/mL≤5 EU/mL≤5 EU/mL≤5 EU/mL
Performance (growth)  *   
9CFR virus testing     
EMA virus testing (selected lots only)     
Biochemical/hormonal profiling     
Filtration     
Total protein (30–45 mg/mL)     
Hemoglobin ≤15 mg/dL≤30 mg/dL≤30 mg/dL≤15 mg/dL
Mycoplasma     
pH (7.0–8.0)     
Osmolality (280–340 mOsm/kgH20)     
Fingerprinting technology (origin confirmation)     
Quality tests per batch96909090
OriginUnited States, Australia, or New Zealand (refer to CoA for specific origin)AustraliaNew ZealandUnited States
Base Cat. Nos.A476610099, 1010010091, 1009316000, 10082, A31604, A38400
Buy 50 mL, 100 mL, 500 mL, or 1 LOrder now †Order nowOrder nowOrder now
Buy 50 mL, 100 mL, 500 mL, or 1 L, heat-inactivatedAvailable upon requestOrder nowAvailable upon requestOrder now
  = testing is performed.
* Additional cell culture proliferation assessment, using 6 widely used cell lines.
† Currently only available in 500 mL bottle.
Gamma-irradiated Premium FBS is available upon request.
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.