KingFisher Duo Prime System
The most versatile compact benchtop sample prep instrument in the lab for 6 or 12 samples per run. Consistent extraction and purification of DNA, RNA, protein, and cells.
Product features
Typical run with KingFisher instrument
KingFisher Flex details
Instrument sizeCompact benchtop
Throughput levelLow to medium
Processing volume range
  • 50–1,000 μL: 12-pin magnet head
  • 200–5,000 μL: 6-pin magnet head
Samples per run6 or 12
Customizable protocolsYes
Heating/cooling
  • 10°C to 75°C (plate row block A)
  • 4°C to 75°C (elution strip block)
Ultraviolet lamp8 watts (up to 16 hr)
How KingFisher instruments work
KingFisher instruments automate extraction of DNA, RNA, protein, and cells by moving magnetic beads (not liquids), leading to clean extractions and enabling consistent results. Using a simple process (bind, wash, elute), KingFisher instruments can automate the extraction of any analyte of interest with a bead on it.
Kits and reagents
Choose from a wide range of magnetic bead-based kits and reagents from trusted brands, such as Dynabeads and MagMAX.
Search now
Plates and plastics
  • Made of polypropylene
  • Low binding affinity for biomolecules, ideal for magnetic particle processing
  • Sterile plastics come with a purity certificate and are guaranteed to be free of RNase, DNase, DNA and endotoxin contamination, shelf life is 5 years
Select different plate formats for a range of input volumes.
 Application note: MagMAX Viral/Pathogen kit protocol changes to enable increased SARS-CoV-2 testing throughput
 KingFisher plastics for SARS-CoV-2 testing
Software and protocols
Nucleic Acid Protocols
  • Protocols designed for nucleic acid purification procedures and reagent kits
Search and download automated protocols
IP/Protein Protocols
  • Protocols for Immunoprecipitation, Protein A, G, Sheep Anti-Mouse, Sheep Anti-Rabbit, Streptavidin
Search and download automated protocols
Learn more
BindIt Software
BindIt Software
  • Create and store protocols in a PC database
  • Compatible with all KingFisher instruments
  • Easy to use
  • Generate reports
Design or customize your own protocols
Service and support
Instrument and application specialists
Whether servicing your KingFisher purification system or addressing your workflow application questions, our more than 1,000 trained professionals make up the industry’s largest network, ready to assist you when you need it.
Quick-start training
The KingFisher system includes SmartStart orientation to get you up and running quickly in your lab. The orientation includes basic familiarization and setup with online instrument management and includes on-site training with standard kits.
Warranty and service plans
Every KingFisher system comes with a one-year warranty. Extended-coverage service plans are also available at the time of instrument purchase. Whether your laboratory requires the highest service levels and adherence to stringent regulatory guidelines, or you need to maximize performance with a limited budget—or anything in between—we’ll work with you to develop a solution that best suits your needs. KingFisher instrument services and support
Find out more at thermofisher.com/instrumentservices or contact us at thermofisher.com/contactus.
Related products for plate setup
To help save time and increase reproducibility with the MagMAX Viral/Pathogen Nucleic Acid Isolation kits, combine with E1-ClipTip Equalizer electronic pipettes and Multidrop Combi instruments.
E1-ClipTip Equalizer Pipettes
E1-ClipTip Equalizer Pipettes
Adjustable spacing, Bluetooth enabled electronic pipette. Learn more
Multidrop Combi Reagent Dispenser
Multidrop Combi Reagent Dispenser
Automated reagent dispenser. Learn more
Product features
Typical run with KingFisher instrument
KingFisher Flex details
Instrument sizeCompact benchtop
Throughput levelLow to medium
Processing volume range
  • 50–1,000 μL: 12-pin magnet head
  • 200–5,000 μL: 6-pin magnet head
Samples per run6 or 12
Customizable protocolsYes
Heating/cooling
  • 10°C to 75°C (plate row block A)
  • 4°C to 75°C (elution strip block)
Ultraviolet lamp8 watts (up to 16 hr)
How KingFisher instruments work
KingFisher instruments automate extraction of DNA, RNA, protein, and cells by moving magnetic beads (not liquids), leading to clean extractions and enabling consistent results. Using a simple process (bind, wash, elute), KingFisher instruments can automate the extraction of any analyte of interest with a bead on it.
Kits and reagents
Choose from a wide range of magnetic bead-based kits and reagents from trusted brands, such as Dynabeads and MagMAX.
Search now
Plates and plastics
  • Made of polypropylene
  • Low binding affinity for biomolecules, ideal for magnetic particle processing
  • Sterile plastics come with a purity certificate and are guaranteed to be free of RNase, DNase, DNA and endotoxin contamination, shelf life is 5 years
Select different plate formats for a range of input volumes.
 Application note: MagMAX Viral/Pathogen kit protocol changes to enable increased SARS-CoV-2 testing throughput
 KingFisher plastics for SARS-CoV-2 testing
Software and protocols
Nucleic Acid Protocols
  • Protocols designed for nucleic acid purification procedures and reagent kits
Search and download automated protocols
IP/Protein Protocols
  • Protocols for Immunoprecipitation, Protein A, G, Sheep Anti-Mouse, Sheep Anti-Rabbit, Streptavidin
Search and download automated protocols
Learn more
BindIt Software
BindIt Software
  • Create and store protocols in a PC database
  • Compatible with all KingFisher instruments
  • Easy to use
  • Generate reports
Design or customize your own protocols
Service and support
Instrument and application specialists
Whether servicing your KingFisher purification system or addressing your workflow application questions, our more than 1,000 trained professionals make up the industry’s largest network, ready to assist you when you need it.
Quick-start training
The KingFisher system includes SmartStart orientation to get you up and running quickly in your lab. The orientation includes basic familiarization and setup with online instrument management and includes on-site training with standard kits.
Warranty and service plans
Every KingFisher system comes with a one-year warranty. Extended-coverage service plans are also available at the time of instrument purchase. Whether your laboratory requires the highest service levels and adherence to stringent regulatory guidelines, or you need to maximize performance with a limited budget—or anything in between—we’ll work with you to develop a solution that best suits your needs. KingFisher instrument services and support
Find out more at thermofisher.com/instrumentservices or contact us at thermofisher.com/contactus.
Related products for plate setup
To help save time and increase reproducibility with the MagMAX Viral/Pathogen Nucleic Acid Isolation kits, combine with E1-ClipTip Equalizer electronic pipettes and Multidrop Combi instruments.
E1-ClipTip Equalizer Pipettes
E1-ClipTip Equalizer Pipettes
Adjustable spacing, Bluetooth enabled electronic pipette. Learn more
Multidrop Combi Reagent Dispenser
Multidrop Combi Reagent Dispenser
Automated reagent dispenser. Learn more