Pacific-TII-Systems
When consistency and convenience are top of mind, choose Thermo Scientific Barnstead Type 2 water purification systems. A wide selection of purification methods, capabilities and options means that you can select the system ideally suited for your specific needs and budget; while low-maintenance operation provides peace-of-mind across your laboratory.
Pure water with a resistivity of >1 Mohm-cm at 25ºC is categorized as Type 2 per ASTM D1193, and is commonly used for general laboratory applications such as buffer, media and reagent preparation, glassware rinsing, and supplying equipment. Type 2 water may also be used as a feed to Type 1 water purification systems.
Interested in distilled water for humidification for lab and nursing home care equipment (ex. oxygen concentrators), dental office autoclaves, and general lab use?  Learn more about our distilled water alternative solution.
Construct a water system for your laboratory requirements
→ Select your water system
→ Choose the product requirements to complete the system
→ Customize your system with optional accessories
→ Request a Quote
Select your water system
Barnstead Type 2 & Dionized Water Systems
Pacific TII
LabTower TII
B-Pure
Harvey DI+
Hose Nipple
Bantam
Deionizer
 product-image-pacificiII
product-image-towerTII
product-image-b-pure
product-image-harveyDI+
product-image-hose-nipple
product-image-bantam-deionizer
DescriptionFlexible installationLarge capacity,
floor mounted
Variety of
cartridge options
Complete systemOptions availableSimple small system
Flow rate
(Model
Dependent)
Up to
7, 12, 20 L/hr
Up to
40 L/hr
Up to
2-4 L/min
Up to
4 L/min
Up to
1.25 L/min
Up to
0.6 L/min
Special
Features
Reverse osmosis and deionization cartridge
Optional
UV lamp
Separate storage
Reverse osmosis and deionization cartridge
Optional
UV lamp
Integrated storage
Deionization cartridge
Double, single,
half size cartridge models
Deionization cartridge
Remote dispenser
Deionization cartridge
Non-pressurized
Deionization cartridge
Non-pressurized
Storage TypesAdjustable storage volumeAdjustable storage volumeNANANANA
Storage
Volume
30, 60,100 L85 L
ControlMicroprocessorMicroprocessor
Digital, resistivity indicatorResistivity indicatorColor indicatorAnalog
MountingBench, wallFreestandingWallWallWallBench, wall
Data Output
Request a quoteOrderOrderOrderOrderOrderOrder
Required accessories
Select the item highlighted in blue for more information.
Optional accessories
Select the item highlighted in blue for more information.
Distilled water is needed for humidification in Oxygen generators and CPAP machines. When water is used for humidification, it is important to have the minerals removed so they do not create deposits or scale in the water reservoir and tubing in Oxygen Concentrators or CPAP machines. This is why distilled water is often recommended as it has the minerals removed from the water.
Harvey DI+ water purification system is an alternative to distilled water.
product-image-harveyDI+
  • The Harvey DI+ is designed to produce purified water by deionization using a resin cartridge which removes the impurities that cause scaling or hard water deposits and can be used as an alternative to distilled water when hard water is a concern.*
  • The system is designed to deliver water on demand vs. purchasing gallon jugs of water.
  • The Harvey DI+ includes a change cartridge light indicator, cartridge and hand dispenser.
  • Applications include distilled water for nursing homes, dentist offices, general lab purposes.
    Download the brochure     or    Order now
* The Harvey DI+ does not make distilled water.
Barnstead Cartridge Distillation-Based Type 2 Water Systems
MegaPure
Classic
 product-image-mega-pure-glass-stills
product-image-classic-stills
DescriptionAll glass stillTin lined still
Production Rate1.4, 3.4, 6 L/hr models1.9, 7.6, 19 L/hr models
Special FeaturesAutomatic Collection System (ACS), Pretreatent options availableQ-Baffle, de-concentrator, optional UV lamp,
optional automatic control, 1.9 L portable unit
Storage Volume6 L plastic,
13 and 45 L glass
94 L tin lined
MountingBench, WallBench (1.9 L),
Floor (7.6, 19 L)
Materials of Contact
Glass, Quartz, PTFE
Copper, Tin
Heat SourceElectricElectric
Request a quote OrderOrder
Required accessories
Select the item highlighted in blue for more information.
MegaPure
Classic
 product-image-mega-pure-glass-stills
product-image-classic-stills
ReservoirsPlastic bottle: 6L ›Tin-lined reservoir: 95L ›
Glass bottle:13L ›
Glass Automatic Collection System (ACS): 45L ›
Floor stands NAReservoir: 95L ›
Air filter and water sealNAVentgard ›
Optional accessories
Select the item highlighted in blue for more information.
MegaPure
Classic
 product-image-mega-pure-glass-still
product-image-classic-stills
Pretreatment
Single cartridge deionizerSingle B-pure ›
NA
High Capacity Cartridge ›
(Recommended)
Adapter Kit › (request information)
Double cartridge deionizerDouble B-pure with 50 Kohm light ›
NA
High Capacity Cartridge ›
(Qty 2 Recommended)
Adapter Kit › (request information)
Dual solenoid Required if using pretreated boiler feedwater and untreated tap cooling waterNA
MP-1 (120 V, MP-1 (240V) ›
MP-3, MP-6A ›
Storage 
Flexible tubing adapter kit (ACS only)
Needed when the supplied glass tubing is either too long or too short to reach still. Flexible tubing is not included.
Flexible tube adaptor › (Recommended)
NA
ACS Wall Mount BracketWall bracket ›NA
ACS AdaptorWasher hook-up for glassware washers  ›NA
Full automatic controlsNAFor use with 7.6 L/hr electric stills (7.6L, 19L) ›
Consumable replacement information
The consumable replacement frequency is recommended below but will depend on the feed water quality and the volume of water processed by the water system. Click on the consumable below or here to learn more ›
Lab Water Source Webinars
Support
  • Unity Lab Services
    Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.