The iBright 1500 Series Imaging Systems were designed to make western blot and gel imaging easy. Click on each subtopic below to learn more.
Applications
The core applications you need and the specialty applications you want
The iBright 1500 Series Imaging Systems offer up to five imaging modes to support your multiple applications. Efficiently and easily capture data from protein gels, nucleic acid gels, chemiluminescent western blots, fluorescent western blots, and more.
Image modeWhat kind of signal can be captured?
Protein gelColorimetric staining of gels (e.g., Coomassie, silver) and membranes (e.g., Ponceau S, Thermo Scientific Pierce Reversible Protein Stain), fluorescent staining of gels (e.g., Invitrogen SYPRO Ruby stain)
Nucleic acid gelEthidium bromide and Invitrogen SYBR stains
Chemiluminescent blotChemiluminescence using all popular HRP and AP substrates (e.g., Thermo Scientific SuperSignal and Invitrogen WesternBreeze substrates)
Fluorescent blotFluorescence with popular RGB (visible range) and near-IR fluorophores (e.g., Invitrogen Alexa Fluor and Alexa Fluor Plus conjugates)
UniversalCustom mode to image samples containing multiple signals, such as chemiluminescence, fluorescence, colorimetric stains, and/or visible images; image display is similar to fluorescent blot mode and allows one to assign false color to any sample
Example imaging applications
Fluorescent western blots
Fluorescent western blots
Chemiluminescent western blots
Chemiluminescent western blots
Combined fluorescent and chemiluminescent western blots
Combined fluorescent and chemiluminescent western blots
Colorimetric western blots
Colorimetric western blots
Fluorescent stained nucleic acid gels
Fluorescent stained nucleic acid gels
Fluorescent stained protein gels
Fluorescent stained protein gels
Colorimetric stained protein gels
Colorimetric stained protein gels
Colorimetric membrane stains
Colorimetric membrane stains
Colony plates
Colony plates
Images pictured for fluorescent stained nucleic acid gels and colorimetric stained protein gels shown in pseudocolor (false color applied). Data is captured in grayscale.
Interface
Load and go—Our 12.1 inch LCD touchscreen interface has a simple, logical layout of functions and features, making our systems easy to use with minimal training. Workflows are similar between imaging modes—for a smooth imaging experience regardless of sample type.
9.1 MP camera
Get publication-quality data fast
ibright-camera-image
Capture crisp, clear, publication-quality images with a 9.1 megapixel cooled CCD camera.
Smart Exposure technology rapidly determines optimal exposure time, minimizing the potential for over- or underexposed images and the need to repeat exposures to get the desired signal.
Automated features
Rather than having to open the sample drawer and repeatedly reposition your sample to achieve proper alignment, iBright 1500 Series Imaging Systems automatically determine the sample position and can rotate samples left or right up to 10° on a mechanically rotating sample stage. Mechanical rotation eliminates the need to digitally rotate the sample, which preserves the integrity of the data, as digital rotation can lead to data alterations.
Zoom function
Digital rotation vs. mechanical rotation. (A) Pixels rotate with digital rotation so bands appear jagged. With mechanical rotation, the sample itself rotates, so bands remain smooth in appearance as the pixels remain aligned. (B) Graphic depicting iBright Imaging System sample stage before and after rotation.
In addition, iBright 1500 Series Imaging Systems automatically determine if the sample requires zoom in order to maximally utilize the 22.5 cm x 18.0 cm field of view. If imaging a single blot, the camera will mechanically move toward the sample up to 2X zoom (8X with additional digital zoom). Mechanical zoom maximizes sensitivity by moving the camera closer to the sample stage and thus reducing focal length. iBright 1500 Series Imaging Systems automatically adjust focus for each level of zoom, producing A crystal-clear images.
Zoom function
Zoom function. (A) Unzoomed image of a fluorescent western blot. (B) Blot at 2X zoom. (C) Blot at 4X zoom. (D) Blot at 8X zoom. (blot not repositioned during successive zooms)
Field of view
Big field of view, small footprint—The iBright Imaging Systems feature a light path design that helps enable a large functional viewing area in a relatively small instrument footprint (68 x 38 x 60cm).
Field of view
The large field of view (22.5 x 18.0 cm) enables capture of up to 4 mini blots or gels.
Filter sets
Accelerate your research—The iBright FL1500 model features five fluorescence channels, permitting up to 4-color fluorescent western blot multiplexing and expanding your possibilities for studying multiple proteins in a single blot. Obtain meaningful and representative comparisons to enhance your experiments. Smart Exposure technology further improves acquisition of multiplex fluorescent western blot data, because signal-to-noise ratios are optimized for each fluorescent channel separately.
Filter sets pre-installed in iBright FL1500 Imaging Systems for visible light range (RGB) and near infrared range (NIR) fluorescent western blotting applications.
Excitation channelFilter range (nm)Emission channelFilter range (nm)Example compatible fluorophores
EX1455-485EM1515-564Alexa Fluor Plus 488, Alexa Fluor 488
EX2515-545EM2568-617Alexa Fluor Plus 555, Alexa Fluor 546
EX3608-632EM3675-720Alexa Fluor Plus 647, Alexa Fluor 594
EX4610-660EM4710-730Alexa Fluor Plus 680, Alexa Fluor 680
EX5745-765EM5800-850Alexa Fluor Plus 800, Alexa Fluor 790
Green-LED transilluminator
Green-LED transilluminator
Go-Green:The iBright Imaging Systems utilize a transilluminator based on green LEDs, which effectively excite popular DNA dyes such ethidium bromide and SYBR Green dye and offer many additional benefits.
No harmful UV rays
No harmful UV rays: While UV light effectively excites many fluorescent dyes and stains, UV light is a health hazard. Further, prolonged exposure to UV light can damage DNA samples, and compromise the integrity of samples to be used for downstream applications, such as subcloning.
No mercury waste
No mercury waste: UV transilluminator bulbs contain mercury, a hazardous substance, and therefore require special care for handling and disposal.
Longer lifetime
Longer lifetime: LED bulbs have a substantially longer real-time life than UV bulbs, which can add up to considerable cost savings over the lifetime of the instrument.