Aliquot
Invitrogen can make aliquots, also called ‘daughter’ plates, for plates that are normalized to either volume or concentration. The minimum volume that our robots can transfer is 10µl and they an only work with 80% of the volume in the normalized plate. For example, the plate is normalized to a volume of 100µl, only 80µl can be used for aliquoting. Please enter the number of aliquots you would like to receive for your plate order and the volume of each oligo sequence.