Plate Size
These are the plate types Invitrogen offers for custom DNA oligos free of charge:
Additionally, we offer two types of 96-well Matrix plates available through our Customer Service team for a fee. Please contact Technical Support for details.
- 96-well plates:
- 200µl: this is a PCR shallow plate (Abgene AB-0800) with a working volume of 180µl. The wells are V-shaped.
- 360µl: this is a standard shallow plate (Corning Costar 3359) with a working volume of 250µl. The shape of the wells is U/round.
- 800µl: this is a standard medium plate (Abgene AB-0765) with a working volume of 650µl. The wells have a conical shape.
- 1200µl: this is a deep well plate (Abgene AB-0564) with a working volume of 800µl. The shape of the wells is U/round
Additionally, we offer two types of 96-well Matrix plates available through our Customer Service team for a fee. Please contact Technical Support for details.
- 384 wells:
- 120µl: this is a standard plate (Abgene AB-0781) with a working volume of 75µl. The shape of the well is pyramidal.
- 65µl: this is a microarray plate (Genetix X7020) with a working volume of 45µl. The bottom of the wells is flat.